(3-(Piperidin-1-yl)phenyl)boronic acid 95%

Supplier: Ambeed
Warning

A117259-100MG A117259-1G A117259-250MG A117259-5G
76822-168EA 19.1 USD
76822-168 76822-170 76822-172 76822-174
(3-(Piperidin-1-yl)phenyl)boronic acid 95%
(3-(Piperidin-1-yl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₂
MW: 205.06 g/mol
MDL Number: MFCD03094966
CAS Number: 634905-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR