Boc-D-2-pyridylalanine 97%

Supplier: Ambeed
Warning

A117116-1G A117116-250MG A117116-5G
76822-154EA 18 USD
76822-154 76822-156 77885-074
Boc-D-2-pyridylalanine 97%
Boc-D-2-pyridylalanine

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₄
MDL Number: MFCD00191191
CAS Number: 98266-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR