7-Methoxy-4H-chromen-4-one 98%

Supplier: Ambeed
Warning

A117059-1G A117059-250MG
76822-150EA 99.73 USD
76822-150 76822-152
7-Methoxy-4H-chromen-4-one 98%
7-Methoxy-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₁₀H₈O₃
MW: 176.169 g/mol
MDL Number: MFCD00218597
CAS Number: 5751-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR