(R)-1,3-Dimethylpiperazine dihydrochloride 97%

Supplier: Ambeed
Warning

A116508-250MG A116508-1G
76822-134EA 37 USD
76822-134 77885-066
(R)-1,3-Dimethylpiperazine dihydrochloride 97%
(R)-1,3-Dimethylpiperazine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₆Cl₂N₂
MDL Number: MFCD12913781
CAS Number: 1152110-26-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR