Ethyl-5-bromo-2-furoate 97%

Supplier: Ambeed

Synonyms: Ethyl 5-bromofuran-2-carboxylate, Ethyl 5-Bromo-2-furoate, Ethyl 5-Bromo-2-furancarboxylate, 5-Bromo-2-furoic acid ethyl ester, 5-Bromo-2-furancarboxylic acid ethyl ester

A116443-25G A116443-5G A116443-1G
76822-130EA 229 USD
76822-130 77570-998 77885-065
Ethyl-5-bromo-2-furoate 97%
Ethyl-5-bromo-2-furoate

Precaución: for Research Use Only

Formula: C₇H₇BrO₃
MDL Number: MFCD00092362
CAS Number: 6132-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR