4-(Chloromethyl)phenylboronic acid pinacol ester 98%

Supplier: Ambeed
Danger

Synonyms: 2-(4-(Chloromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A116202-1G A116202-100MG A116202-250MG
76822-110EA 88 USD
76822-110 77885-062 77885-063
4-(Chloromethyl)phenylboronic acid pinacol ester 98%
4-(Chloromethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BClO₂
MDL Number: MFCD11053893
CAS Number: 1072945-04-0
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR