3-Bromo-2-chloro-6-methoxypyridine 98%

Supplier: Ambeed
Warning

A115943-1G A115943-250MG A115943-5G
76822-102EA 82.76 USD
76822-102 76822-104 76822-106
3-Bromo-2-chloro-6-methoxypyridine 98%
3-Bromo-2-chloro-6-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClNO
MDL Number: MFCD10699710
CAS Number: 777931-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR