1-(3-Fluoropyridin-4-yl)ethanone 98%

Supplier: Ambeed
Warning

A115722-100MG A115722-10G A115722-1G A115722-250MG A115722-5G
76822-092EA 24 USD
76822-092 76822-094 76822-096 76822-098 76822-100
1-(3-Fluoropyridin-4-yl)ethanone 98%
1-(3-Fluoropyridin-4-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₆FNO
MDL Number: MFCD07375077
CAS Number: 87674-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR