4-(Ethoxymethyl)phenol 97%

Supplier: Ambeed
Warning

A115679-1G A115679-250MG
76822-088EA 682 USD
76822-088 76822-090
4-(Ethoxymethyl)phenol 97%
4-(Ethoxymethyl)phenol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MDL Number: MFCD00464284
CAS Number: 57726-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR