Ethyl 2-bromo-5-fluorobenzoate 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A115165-100G A115165-10G A115165-25G A115165-5G A115165-1G
76822-064EA 173.99 USD
76822-064 76822-066 76822-068 76822-070 77683-048
Ethyl 2-bromo-5-fluorobenzoate 97%, Ambeed.Inc
Ethyl 2-bromo-5-fluorobenzoate

Precaución: For research use only.

Formula: C₉H₈BrFO₂
MW: 247.061 g/mol
MDL Number: MFCD06204688
CAS Number: 139911-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR