2,6-Dimethoxyisonicotinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dimethoxypyridine-4-carboxylic acid

A114452-250MG A114452-100MG
76822-048EA 84.88 USD
76822-048 77683-039
2,6-Dimethoxyisonicotinic acid 98%
2,6-Dimethoxyisonicotinic acid

Precaución: For research use only.

Formula: C₈H₉NO₄
MW: 183.17 g/mol
MDL Number: MFCD18257556
CAS Number: 6274-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR