(2R,3S,5R)-5-(6-Benzamido-9H-purin-9-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite 95%

Supplier: Ambeed
Warning

A114451-10G A114451-1G A114451-250MG A114451-5G A114451-25G A114451-100G
76822-040EA 75 USD
76822-040 76822-042 76822-044 76822-046 77570-812 77885-054
(2R,3S,5R)-5-(6-Benzamido-9H-purin-9-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite 95%
(2R,3S,5R)-5-(6-Benzamido-9H-purin-9-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite

Precaución: for Research Use Only

Formula: C₄₇H₅₂N₇O₇P
MDL Number: MFCD00036314
CAS Number: 98796-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR