2,7-Dichlorobenzo[d]thiazole 97%

Supplier: Ambeed
Warning

A114268-100MG A114268-1G A114268-250MG
76822-022EA 20.16 USD
76822-022 76822-024 76822-026
2,7-Dichlorobenzo[d]thiazole 97%
2,7-Dichlorobenzo[d]thiazole

Precaución: for Research Use Only

Formula: C₇H₃Cl₂NS
MW: 204.08 g/mol
MDL Number: MFCD09037778
CAS Number: 2942-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR