Ethyl 1-methyl-5-nitro-1H-imidazole-2-carboxylate 98%

Supplier: Ambeed
Warning

A114210-1G A114210-250MG A114210-5G
76822-008EA 124.13 USD
76822-008 76822-010 76822-012
Ethyl 1-methyl-5-nitro-1H-imidazole-2-carboxylate 98%
Ethyl 1-methyl-5-nitro-1H-imidazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉N₃O₄
MW: 199.16 g/mol
MDL Number: MFCD03789116
CAS Number: 1564-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR