8-Bromo-6-chloro-2-methyl-[1,2,4]triazolo[1,5-a]pyridine 98%

Supplier: Ambeed
Warning

A114202-100MG A114202-1G A114202-250MG
76822-002EA 41.38 USD
76822-002 76822-004 76822-006
8-Bromo-6-chloro-2-methyl-[1,2,4]triazolo[1,5-a]pyridine 98%
8-Bromo-6-chloro-2-methyl-[1,2,4]triazolo[1,5-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₅BrClN₃
MW: 246.49 g/mol
MDL Number: MFCD12400879
CAS Number: 1159813-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR