9-(2-Bromophenyl)-9H-carbazole 97%
Supplier: Ambeed
|
Warning
|
A113678-10G
A113678-1G
A113678-250MG
A113678-25G
A113678-5G
A113678-100G
76821-986EA
37.14
USD
76821-986
76821-988
76821-990
76821-992
76821-994
77683-035
9-(2-Bromophenyl)-9H-carbazole 97%
9-(2-Bromophenyl)-9H-carbazole
Precaución: For research use only.
Formula:
C₁₈H₁₂BrN MW: 322.21 g/mol |
MDL Number:
MFCD23135883 CAS Number: 902518-11-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...