5-(1-Propynyl)pyridine-3-boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (5-(Prop-1-yn-1-yl)pyridin-3-yl)boronic acid

A113452-100MG A113452-1G A113452-250MG
76821-980EA 42.44 USD
76821-980 76821-982 76821-984
5-(1-Propynyl)pyridine-3-boronic acid 97%
5-(1-Propynyl)pyridine-3-boronic acid

Precaución: for Research Use Only

Formula: C₈H₈BNO₂
MW: 160.97 g/mol
MDL Number: MFCD18258851
CAS Number: 917471-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR