3-Iodo-1H-indazole-5-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A113420-250MG
A113420-1G
A113420-5G
76821-978EA
9
USD
76821-978
77885-051
77885-052
3-Iodo-1H-indazole-5-carboxylic acid 95%
3-Iodo-1H-indazole-5-carboxylic acid
Precaución: for Research Use Only
Formula:
C₈H₅IN₂O₂ |
MDL Number:
MFCD07781585 CAS Number: 885521-46-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...