2-(Aminomethyl)-5-(trifluoromethyl)pyridine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 5-(Trifluoromethyl)picolinylamine hydrochloride, [5-(Trifluoromethyl)pyridin-2-yl]methylamine hydrochloride, (5-(Trifluoromethyl)pyridin-2-yl)methanamine hydrochloride

A113245-1G A113245-250MG
76821-968EA 99.73 USD
76821-968 76821-970
2-(Aminomethyl)-5-(trifluoromethyl)pyridine hydrochloride 95%
2-(Aminomethyl)-5-(trifluoromethyl)pyridine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₇F₃N₂·ClH
MW: 212.6 g/mol
MDL Number: MFCD05863878
CAS Number: 871826-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR