[3-(4-Fluorophenyl)-5-methylisoxazol-4-yl]methanol 97%

Supplier: Ambeed
Warning

A112830-1G A112830-250MG
76821-946EA 171 USD
76821-946 76821-948
[3-(4-Fluorophenyl)-5-methylisoxazol-4-yl]methanol 97%
[3-(4-Fluorophenyl)-5-methylisoxazol-4-yl]methanol

Precaución: for Research Use Only

Formula: C₁₁H₁₀FNO₂
MDL Number: MFCD10008830
CAS Number: 1018297-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR