(3R,5S)-rel-tert-Butyl 3,5-dimethylpiperazine-1-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A112440-100MG
A112440-10G
A112440-1G
A112440-250MG
A112440-25G
A112440-5G
76821-922EA
5
USD
76821-922
76821-924
76821-926
76821-928
76821-930
76821-932
(3R,5S)-rel-tert-Butyl 3,5-dimethylpiperazine-1-carboxylate 98%
(3R,5S)-rel-tert-Butyl 3,5-dimethylpiperazine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₁H₂₂N₂O₂ |
MDL Number:
MFCD06795911 CAS Number: 129779-30-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...