6-Aminonicotinaldehyde hydrochloride 97%

Supplier: Ambeed
Warning

A112288-100MG A112288-1G A112288-250MG
76821-910EA 5 USD
76821-910 76821-912 76821-914
6-Aminonicotinaldehyde hydrochloride 97%
6-Aminonicotinaldehyde hydrochloride

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O
MW: 158.58 g/mol
MDL Number: MFCD22570335
CAS Number: 1588441-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR