2-Bromocarbazole 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-9H-carbazole

A112061-100G A112061-10G A112061-1G A112061-250MG A112061-25G A112061-5G
76821-886EA 149 USD
76821-886 76821-888 76821-890 76821-892 76821-894 76821-896
2-Bromocarbazole 98%
2-Bromocarbazole

Precaución: for Research Use Only

Formula: C₁₂H₈BrN
MDL Number: MFCD09750430
CAS Number: 3652-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR