2,3-Dichloro-4-pyridineboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dichloropyridine-4-boronic acid, pinacol ester, 2,3-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A111660-250MG A111660-10G A111660-1G
76821-872EA 13 USD
76821-872 77885-028 77885-029
2,3-Dichloro-4-pyridineboronic acid pinacol ester 97%
2,3-Dichloro-4-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₄BCl₂NO₂
MDL Number: MFCD06798257
CAS Number: 1073353-78-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR