Methyl 6-oxospiro[3.3]heptane-2-carboxylate 95%

Supplier: Ambeed
Warning

A111347-100MG A111347-1G A111347-250MG A111347-5G A111347-25G
76821-854EA 35.01 USD
76821-854 76821-856 76821-858 76821-860 77683-019
Methyl 6-oxospiro[3.3]heptane-2-carboxylate 95%
Methyl 6-oxospiro[3.3]heptane-2-carboxylate

Precaución: For research use only.

Formula: C₉H₁₂O₃
MW: 168.19 g/mol
MDL Number: MFCD20922770
CAS Number: 1138480-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR