Methyl 4-hydroxy-3,5-diiodobenzoate 97%
Supplier: Ambeed
|
Warning
|
A110980-1G
A110980-250MG
A110980-25G
76821-820EA
16
USD
76821-820
76821-822
77570-626
Methyl 4-hydroxy-3,5-diiodobenzoate 97%
Methyl 4-hydroxy-3,5-diiodobenzoate
Precaución: for Research Use Only
Formula:
C₈H₆I₂O₃ |
MDL Number:
MFCD03425425 CAS Number: 3337-66-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...