4-Chloro-[1,1'-biphenyl]-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A110784-100MG A110784-1G A110784-250MG A110784-5G
76821-802EA 35.01 USD
76821-802 76821-804 76821-806 76821-808
4-Chloro-[1,1'-biphenyl]-3-carboxylic acid 95%
4-Chloro-[1,1'-biphenyl]-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₉ClO₂
MW: 232.66 g/mol
MDL Number: MFCD09042509
CAS Number: 855207-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR