L-Threonine 97%

Supplier: Ambeed
Warning

A110778-1000G A110778-100G A110778-500G
76821-794EA 33 USD
76821-794 76821-796 76821-798 76821-800
L-Threonine 97%
L-Threonine

Precaución: for Research Use Only

Formula: C₄H₉NO₃
MDL Number: MFCD00064270
CAS Number: 72-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR