5-Amino-piperidin-2-one hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 5-Aminopiperidin-2-one hydrochloride

A110677-100MG A110677-1G
76821-784EA 81.69 USD
76821-784 76821-786
5-Amino-piperidin-2-one hydrochloride 95%
5-Amino-piperidin-2-one hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁ClN₂O
MW: 150.61 g/mol
MDL Number: MFCD09743647
CAS Number: 1235440-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR