4-Fluoro-4-Piperidinemethanol hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (4-Fluoropiperidin-4-yl)methanol hydrochloride

A110297-100MG A110297-250MG
76821-764EA 35.01 USD
76821-764 76821-766
4-Fluoro-4-Piperidinemethanol hydrochloride 97%
4-Fluoro-4-Piperidinemethanol hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂FNO·ClH
MW: 169.625 g/mol
MDL Number: MFCD19105116
CAS Number: 1254115-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR