Potassium(3-pyridyl)trifluoroborate 97%

Supplier: Ambeed
Warning

A109809-1G A109809-25G
76821-742EA 22 USD
76821-742 77885-004
Potassium(3-pyridyl)trifluoroborate 97%
Potassium (3-pyridyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₅H₄BF₃KN
MDL Number: MFCD09038545
CAS Number: 561328-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR