5-Bromo-6-chloro-3-nitropyridin-2-amine 96%
Supplier: Ambeed
|
Warning
|
A109504-1G
A109504-250MG
A109504-5G
76821-722EA
43.5
USD
76821-722
76821-724
76821-726
5-Bromo-6-chloro-3-nitropyridin-2-amine 96%
5-Bromo-6-chloro-3-nitropyridin-2-amine
Precaución: for Research Use Only
Formula:
C₅H₃BrClN₃O₂ MW: 252.45 g/mol |
MDL Number:
MFCD20230539 CAS Number: 1335057-22-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...