(1-Methyl-1H-indazol-5-yl)methanol 95%

Supplier: Ambeed
Warning

A109408-250MG A109408-5G A109408-1G
76821-712EA 34 USD
76821-712 76821-714 77884-996
(1-Methyl-1H-indazol-5-yl)methanol 95%
(1-Methyl-1H-indazol-5-yl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O
MDL Number: MFCD11052640
CAS Number: 1092961-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR