2-(2-Bromo-5-methoxyphenyl)acetonitrile 97%

Supplier: Ambeed
Warning

A109381-100MG A109381-1G A109381-250MG
76821-706EA 24.41 USD
76821-706 76821-708 76821-710
2-(2-Bromo-5-methoxyphenyl)acetonitrile 97%
2-(2-Bromo-5-methoxyphenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₉H₈BrNO
MW: 226.07 g/mol
MDL Number: MFCD09927439
CAS Number: 27387-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR