1,4,7,10-Tetrakis(aminocarbonylmethyl)-1,4,7,10-tetraazacyclododecane 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
DOTA-amide, 2,2',2'',2'''-(1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetrayl)tetraacetamide
A108491-100MG
A108491-1G
A108491-250MG
A108491-10G
A108491-5G
76821-672EA
12.74
USD
76821-672
76821-674
76821-676
77682-995
77682-996
1,4,7,10-Tetrakis(aminocarbonylmethyl)-1,4,7,10-tetraazacyclododecane 97%
1,4,7,10-Tetrakis(aminocarbonylmethyl)-1,4,7,10-tetraazacyclododecane
Precaución: For research use only.
Formula:
C₁₆H₃₂N₈O₄ MW: 400.48 g/mol |
MDL Number:
MFCD09263318 CAS Number: 157599-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...