(4-Chloro-2-methylsulfanylpyrimidin-5-yl)methanol 97%

Supplier: Ambeed
Warning

Synonyms: (4-Chloro-2-(methylthio)pyrimidin-5-yl)methanol

A108469-100MG A108469-250MG
76821-668EA 6.37 USD
76821-668 76821-670
(4-Chloro-2-methylsulfanylpyrimidin-5-yl)methanol 97%
(4-Chloro-2-methylsulfanylpyrimidin-5-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₇ClN₂OS
MW: 190.65 g/mol
MDL Number: MFCD11858523
CAS Number: 1044145-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR