Methyl 3-bromo-4-formylbenzoate 98%

Supplier: Ambeed
Warning

A108040-100MG A108040-1G A108040-250MG A108040-5G
76821-656EA 29.71 USD
76821-656 76821-658 76821-660 76821-662
Methyl 3-bromo-4-formylbenzoate 98%
Methyl 3-bromo-4-formylbenzoate

Precaución: for Research Use Only

Formula: C₉H₇BrO₃
MW: 243.054 g/mol
MDL Number: MFCD16036864
CAS Number: 90484-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR