2-Ethylpentanoic acid 95%, Ambeed.Inc

Supplier: Ambeed
Danger

A107733-1G A107733-100MG A107733-5G A107733-250MG
76821-642EA 90 USD
76821-642 77682-987 77682-988 77884-979
2-Ethylpentanoic acid 95%, Ambeed.Inc
2-Ethylpentanoic acid

Precaución: For research use only.

Formula: C₇H₁₄O₂
MDL Number: MFCD02258650
CAS Number: 20225-24-5
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR