2-Mercaptobenzo[d]thiazole-6-carbonitrile 96%

Supplier: Ambeed
Warning

A107565-100MG A107565-1G A107565-250MG
76821-624EA 51.99 USD
76821-624 76821-626 76821-628
2-Mercaptobenzo[d]thiazole-6-carbonitrile 96%
2-Mercaptobenzo[d]thiazole-6-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₄N₂S₂
MW: 192.26 g/mol
MDL Number: MFCD11845316
CAS Number: 315228-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR