2-Amino-9,9-diphenylfluorene 97%

Supplier: Ambeed
Warning

A107374-10G A107374-1G A107374-250MG A107374-25G
76821-610EA 61 USD
76821-610 76821-612 76821-614 77570-448
2-Amino-9,9-diphenylfluorene 97%
2-Amino-9,9-diphenylfluorene

Precaución: for Research Use Only

Formula: C₂₅H₁₉N
MDL Number: MFCD22376664
CAS Number: 1268519-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR