5-Bromo-4-methoxy-2-methylaniline 97%

Supplier: Ambeed
Warning

A107187-250MG A107187-5G A107187-1G
76821-606EA 14 USD
76821-606 76821-608 77884-977
5-Bromo-4-methoxy-2-methylaniline 97%
5-Bromo-4-methoxy-2-methylaniline

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO
MDL Number: MFCD17392823
CAS Number: 861084-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR