BAY 11-7082 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A107077-10MG A107077-1MG A107077-250MG A107077-50MG A107077-5MG A107077-1G A107077-100MG
76821-596EA 40 USD
76821-596 76821-598 76821-600 76821-602 76821-604 77682-984 77884-976
BAY 11-7082 98%, Ambeed.Inc
BAY 11-7082

Precaución: For research use only.

Formula: C₁₀H₉NO₂S
MDL Number: MFCD00712162
CAS Number: 19542-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR