tert-Butyl 3-amino-1H-indazole-1-carboxylate 95%

Supplier: Ambeed
Warning

A106725-1G A106725-250MG
76821-592EA 467.86 USD
76821-592 76821-594
tert-Butyl 3-amino-1H-indazole-1-carboxylate 95%
tert-Butyl 3-amino-1H-indazole-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₅N₃O₂
MW: 233.26 g/mol
MDL Number: MFCD12827767
CAS Number: 1204298-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR