tert-Butyl 4-(4-amino-2-cyanophenyl)piperazine-1-carboxylate 97%

Supplier: Ambeed
Warning

A106469-100MG A106469-250MG A106469-5G
76821-572EA 27.59 USD
76821-572 76821-574 76821-576
tert-Butyl 4-(4-amino-2-cyanophenyl)piperazine-1-carboxylate 97%
tert-Butyl 4-(4-amino-2-cyanophenyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₆H₂₂N₄O₂
MW: 302.37 g/mol
MDL Number: MFCD23135819
CAS Number: 288251-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR