2-Chloro-7-nitroquinoline 97%

Supplier: Ambeed
Warning

A106162-100MG A106162-250MG A106162-1G A106162-5G
76821-566EA 91 USD
76821-566 76821-568 77565-854 77884-966
2-Chloro-7-nitroquinoline 97%
2-Chloro-7-nitroquinoline

Precaución: For research use only.

Formula: C₉H₅ClN₂O₂
MDL Number: MFCD18448576
CAS Number: 49609-03-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR