[(5-Chloro-1H-indol-2-yl)methyl]amine 95%

Supplier: Ambeed
Warning

A106065-100MG A106065-1G
76821-564EA 68 USD
76821-564 77884-965
[(5-Chloro-1H-indol-2-yl)methyl]amine 95%
[(5-Chloro-1H-indol-2-yl)methyl]amine

Precaución: for Research Use Only

Formula: C₉H₉ClN₂
MDL Number: MFCD02089399
CAS Number: 21109-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR