4,5-Difluoro-2-iodoaniline 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A106013-250MG A106013-1G A106013-5G A106013-25G
76821-562EA 14 USD
76821-562 77682-976 77884-964 77884-963
4,5-Difluoro-2-iodoaniline 95%, Ambeed.Inc
4,5-Difluoro-2-iodoaniline

Precaución: For research use only.

Formula: C₆H₄F₂IN
MDL Number: MFCD03095321
CAS Number: 847685-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR