4-Bromo-1-chloro-2-(chloromethyl)benzene 95%

Supplier: Ambeed
Danger

A105878-1G A105878-5G
76821-548EA 24.41 USD
76821-548 76821-550
4-Bromo-1-chloro-2-(chloromethyl)benzene 95%
4-Bromo-1-chloro-2-(chloromethyl)benzene

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂
MW: 239.92 g/mol
MDL Number: MFCD12024979
CAS Number: 928758-19-4
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR