2,4-Dibromoiodobenzene 97%

Supplier: Ambeed
Warning

Synonyms: 2,4-Dibromo-1-iodobenzene

A105861-1G A105861-25G A105861-5G A105861-100G
76821-542EA 13.8 USD
76821-542 76821-544 76821-546 77682-974
2,4-Dibromoiodobenzene 97%
2,4-Dibromoiodobenzene

Precaución: For research use only.

Formula: C₆H₃Br₂I
MW: 361.8000 g/mol
MDL Number: MFCD09753709
CAS Number: 19393-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR