1,3,5-Tris(4-iodophenyl)benzene 98%

Supplier: Ambeed
Warning

Synonyms: 4,4''-Diiodo-5'-(4-iodophenyl)-1,1':3',1''-terphenyl

A105403-10G A105403-1G A105403-250MG A105403-5G A105403-100MG
76821-528EA 171 USD
76821-528 76821-530 76821-532 76821-534 77884-959
1,3,5-Tris(4-iodophenyl)benzene 98%
1,3,5-Tris(4-iodophenyl)benzene

Precaución: for Research Use Only

Formula: C₂₄H₁₅I₃
MW: 684.10 g/mol
MDL Number: MFCD07369795
CAS Number: 151417-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR